Orders LinkContactUsLinkComponent
Modified EC Medium, CRITERION™, Hardy Diagnostics
Modified EC Medium, CRITERION™, Hardy Diagnostics
Modified EC Medium, CRITERION™, Hardy Diagnostics
Certificates
Modified EC Medium, CRITERION™, Hardy Diagnostics
Modified EC Medium, CRITERION™, Hardy Diagnostics
1 Options Available Below
Product Details & Documents
CRITERION™ Modified EC Medium is for the detection of Escherichia coli 0157:H
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA89406-678
  • CA89406-680
  • CA89406-682
  • CA89406-684
  • Storage temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Final volume
    Pack type
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-678
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $484.00
    $484.00
    Each
       $484.00
    Minimum order quantity is 1
    Supplier Number: C8091
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-680
    Storage temperature
    –2-30°C
  • 2 kg
    Bucket
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-682
    Storage temperature
    –2-30°C
  • 10 kg
    Bucket
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-684
    Storage temperature
    –2-30°C