Orders LinkContactUsLinkComponent
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Certificates
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
4 Options Available Below
Product Details & Documents
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
 : BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA89405-362
  • CA89405-364
  • CA89405-366
  • CA89405-368
  • Storage temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Final volume
    Pack type
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Log in for availability
    Log in for availability
    Each
    Each - $261.00
    $261.00
    Each
       $261.00
    Supplier Number: C5570
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-362
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $934.00
    $934.00
    Each
       $934.00
    Supplier Number: C5571
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-364
    Storage temperature
    –2-30°C
  • 2 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $3,680.00
    $3,680.00
    Each
       $3,680.00
    Supplier Number: C5572
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-366
    Storage temperature
    –2-30°C
  • 10 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $18,440.00
    $18,440.00
    Each
       $18,440.00
    Supplier Number: C5573
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-368
    Storage temperature
    –2-30°C