CRITERION™ Agar, Pharmaceutical Grade, Hardy Diagnostics
About this item
CRITERION™ Agar, Pharmaceutical Grade is primarily a solidifying agent which is purified to remove extraneous matter, pigments and salts.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
Pharmaceutical grade is suitable for use in preparing culture media. Its wide mouth opening reduces dust and allows for easy scooping. Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free
3 Options Available Below
Product Details & Documents
CRITERION™ Agar, Pharmaceutical Grade is primarily a solidifying agent which is purified to remove extraneous matter, pigments and salts.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
Pharmaceutical grade is suitable for use in preparing culture media. Its wide mouth opening reduces dust and allows for easy scooping. Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89406-342
- CA89406-344
- CA89406-346
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: C7431Specifications:
More
Specifications
Cat. No.CA89406-342Storage temperature–2-30°C - Supplier Number: C7432Specifications:
More
Specifications
Cat. No.CA89406-344Storage temperature–2-30°C - Supplier Number: C7433Specifications:
More
Specifications
Cat. No.CA89406-346Storage temperature–2-30°C