Heating Tapes, Lab Type, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Glas-Col heating tapes allow even short tapes, such as those 0.9m long, to be used on 115 V without the need for a reducing voltage transformer.
They can be operated directly through a temperature controller at normal outlet voltage. Operating temperature up to 249 °C (480 °F). Both power supply connections are made at one end of the tape. All tapes are supplied with separable 1.2 m extension cord with two-blade cup. Supplied complete with cord and plug.
Customers Who Bought This Also Bought