ContactUsLinkComponent
Compact Extra-Wide Non-Metal Locking Carts, 4-Drawer Polyethylene Polymer, TrippNT
Compact Extra-Wide Non-Metal Locking Carts, 4-Drawer Polyethylene Polymer, TrippNT
Supplier:  TrippNT
Certificates
undefined
Compact Extra-Wide Non-Metal Locking Carts, 4-Drawer Polyethylene Polymer, TrippNT
Supplier:  TrippNT
Specifications
  • Catalog No:
  • 10805-268
  • Material:
  • Food Grade High Density Polyethylene, Natural White; ABS (Acrylonitrile butadiene styrene) drawers; PVC drawer fronts
  • Storage capacity:
  • 4.7 cu. ft.
  • Caster type:
  • 3" total locking polypropylene casters (locks rotation and swivel)
  • Drawer dimensions:
  • 24½W×4½H×15½" D
  • Tray dimensions:
  • 25¼×3¼×15⅝", top
  • Width:
  • 25⅜"
  • Depth:
  • 18⅛"
  • Height:
  • 27⅝"
  • Weight:
  • 67 lbs.