Order Entry
Canada
ContactUsLinkComponent
MRI Core DX Extended Top Storage Cart, Polyethylene, TrippNT
MRI Core DX Extended Top Storage Cart, Polyethylene, TrippNT
Catalog # 89408-478
Supplier:  TrippNT
undefined
MRI Core DX Extended Top Storage Cart, Polyethylene, TrippNT
Catalog # 89408-478
Supplier:  TrippNT
Supplier Number:  51817

Specifications

  • Color:
    White with White Drawer and Clear Door
  • Description:
    MRI Core DX Extended Top Storage Cart
  • Cat. no.:
    89408-478
  • Weight:
    62 lbs.
  • Supplier No.:
    51817

Specifications

About this item

Non ferrous, white polyethylene MRI Core DX Extended Top Lab Cart provides flexible, extra storage and workspace with an extra large 29 x 19 inch work surface.

  • Non-ferrous MRI organizational cart
  • X-Wide flat top work surface
  • 11 flexible storage features
  • 3 drawers
  • 6 side shelves
  • Bulk storage area
  • Tool holder
  • Chemically resistant polyethylene construction
  • Top has two built in handles making it smooth to maneuver
  • 3 inch casters
  • Fully assembled, ready to use out of the box
  • In stock and ready to ship
  • American made by TrippNT
  • Total Replacement Lifetime Guarantee

Made of sturdy polyethylene and ABS plastic construction. Push and pull with two comfortable built-in handles.

It features 3 inch, non-marking rubber casters, 6 side shelves, 3 full extension drawers, 2 glove dispensers, and a bulk storage area. These MR Conditional carts add flexible storage for mobility and efficiency.