Polywipe-C 'Blue Inspectr' Wipes, Contec®
Polywipe-C "Blue InspectR" Wipes, 75/Inner Bag, 2 Inner Bags/Outer Bag, 12 Outer Bags/Case, 22.9×22.9 cm (9×9")
Supplier #: PCB-99
Polywipe-C 'Blue Inspectr' Wipes, Contec®
Catalog # 33501-476
Small Business Enterprise
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WIPE KNIT-LITE 9X9IN PK150 Catalog #68310-160
Product 68310-160 is similar to 33501-476 but manufacturer and specifications may vary.
Product Details & Documents
Polywipe-C 'Blue InspectR' wipes are specifically designed to reveal particle contaminants picked-up from wiped surfaces.
- Dark blue color reveals light-colored contaminants
- Good sorbency with solvents
- Blue color-fast dye has been applied to provide visual contrast without added extractables
The fabric is lightweight and has a textured surface for effective cleaning without scratching. The wipe fabric is solution dyed to provide visual contrast without added extractables. The dye is colorfast and will not bleed when used with most solutions and solvents.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Polywipe-C "Blue InspectR" Wipes
- Packaging:75/Inner Bag, 2 Inner Bags/Outer Bag, 12 Outer Bags/Case
- W×L:22.9×22.9 cm (9×9")
- Cat. No.:33501-476
- Material:Polyester knit



