To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Regulatory status:RUO
- Cat. no.:CACB355006
- Shelf Life:790 days
- Volume:5 mL
- Supplier No.:355006
Specifications
About this item
MITO+ Serum Extender is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steriod hormones)
It can be used to culture a variety of cells under serum-free or serum-reduced conditions.
Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.
(5ml; 5L equivalent).