To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Rat
- Regulatory status:RUO
- Cat. no.:CACB354115
- Source:Rat splenocyte cultures
- Shelf Life:790 days
- Volume:100 mL
- Supplier No.:354115
Specifications
About this item
T-STIM with ConA (IL-2 culture supplement), rat, is used to promote proliferation and activation of T-cells and support high-titer HIV production by leukocytes.
Media used to promote proliferation and activation (PHA or ConA Supplemented Media) of T-cells and support high-titer HIV production by leukocytes.
Tested for ability to promote proliferation of an IL-2 dependent mouse cell line (CTLL) in serum-containing medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.