Corning® Basic Fibroblast Growth Factor (bFGF), Corning
10 µg
Supplier: Corning
Supplier #: 354060
Corning® Basic Fibroblast Growth Factor (bFGF), Corning
Catalog # CACB354060
Supplier: Corning
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Basic Fibroblast Growth Factors (bFGF), human recombinant, are heparin-binding mitogenic proteins that enhance proliferation of a wide variety of cell types under serum-free or serum-reduced conditions.
- Growth Factor
Can also be used as a chemotactic or neurotrophic factor and to study wound healing, angiogenesis, and related processes.
Tested for ability to promote proliferation of fetal bovine heart endothelial cells (FBHEC).
Caution: Regulatory status: RUO
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Size:10 µg
- Cat. No.:CACB354060
- Protein/peptide type:Recombinant
- Protein/peptide name:bFGF
- Protein synonyms:Basic Fibroblast Growth Factor
- Purity:Purity > 97%by SDS-PAGE
- Species:Human
- Source:E. coli
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



