ContactUsLinkComponent
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Catalog # 89166-062
Supplier:  Enzo Life Sciences
CAS Number:  
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Catalog # 89166-062
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-53003-M010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mito-Id Red Antifade Reagent
  • Size:
    10 ml
  • Environmentally Preferable:
  • Cat. no.:
    89166-062
  • Supplier No.:
    53003-M010
Specifications
About this item
Mitochondria dye.
Developed specifically for Mito-ID™ Red mitochondrial stain, but works well with many other stains, such as fluorescein and rhodamine. Causes little or no quenching.
Use of the Mito-ID™ Red Antifade Reagent with DAPI with the Mito-ID™ Red dye, or other stains, significantly extends the photobleaching half-life of the fluorescent stain. The addition of DAPI to the mounting medium allows for staining of the nucleus.