Order Entry
Canada
ContactUsLinkComponent
PROTEOSTAT® PDI Assay Kit, Enzo Life Sciences
PROTEOSTAT® PDI Assay Kit, Enzo Life Sciences
Catalog # 89165-912
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
PROTEOSTAT® PDI Assay Kit, Enzo Life Sciences
Catalog # 89165-912
Supplier:  Enzo Life Sciences
Supplier Number:  51024-KP002
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    2x96 Tests
  • Environmentally Preferable:
  • Storage conditions:
    –80 °C (Long Term)
  • Application:
    Fluorescent Detection
  • Cat. no.:
    89165-912
  • Supplier No.:
    51024-KP002

Specifications

About this item

The PROTEOSTAT® PDI assay kit provides a simple, homogenous assay for screening modulators of PDI enzymatic activity.

The PROTEOSTAT® PDI assay kit is capable of providing a quantitative readout of PDI enzymatic activity in a robust and high-throughput fashion and can be applied to identification of PDI inhibitors from chemical libraries.

This is accomplished by monitoring PDI-catalyzed reduction of insulin in the presence of Dithiothreitol (DTT), resulting in the formation of insulin aggregates which then bind avidly to the red-emitting fluorogenic PROTEOSTAT® PDI detection dye.

Delivery: Kit includes PROTEOSTAT® PDI detection reagent, PDI (human) (recombinant), Insulin (Lyophilized), Bacitracin (Inhibitor Control, Lyophilized), PBE buffer, Stop reagent, DTT (10 mM), Deionized water.