Order Entry
Canada
ContactUsLinkComponent
ROS-ID® Superoxide Detection Kit, Enzo Life Sciences
ROS-ID® Superoxide Detection Kit, Enzo Life Sciences
Catalog # 89165-886
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
ROS-ID® Superoxide Detection Kit, Enzo Life Sciences
Catalog # 89165-886
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-51012
CAS Number:  

Specifications

  • Size:
    1 Kit
  • Application:
    Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection and HTS
  • Cat. no.:
    89165-886
  • Supplier No.:
    51012

Specifications

About this item

This kit is designed to directly monitor real time superoxide production in live cells using fluorescence microscopy and/or flow cytometry.

A major component of the kit, Superoxide Detection Reagent (Orange), is a cell-permeable probe that reacts specifically with superoxide, generating an orange fluorescent product.

The kit is not designed to detect reactive peroxide, hydroxyl, peroxynitrite, chlorine or bromine species, as the fluorescent probe included is relatively insensitive to these analytes. Upon staining, the fluorescent product generated can be visualized using a wide-field fluorescence microscope equipped with a standard orange (e.g., 550/620 nm) fluorescent cube, or cytometrically using any flow cytometer equipped with a blue (488 nm) laser.

The Superoxide Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in suspension).

Delivery: Superoxide Detection Reagent (Orange), 300 nmoles, ROS Inducer (Pyocyanin), 1 µmole, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and Wash Buffer Salts, 1 pack.