Specifications
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection and HTS
- Cat. no.:89165-884
- Supplier No.:51011
Specifications
About this item
This kit is designed to directly monitor real time reactive oxygen and/or nitrogen species (ROS/RNS) production in live cells using fluorescence microscopy and/or flow cytometry.
The kit includes Oxidative Stress Detection Reagent (Green) as the major component
The kit is not designed to detect superoxide, reactive chlorine or bromine species, as the fluorescent probe included is relatively insensitive to these analytes. Upon staining, the fluorescent product generated can be visualized using a wide-field fluorescence microscope equipped with standard green filter (490/525 nm), or cytometrically using any flow cytometer equipped with a blue (488 nm) laser.
The Total ROS Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in solution).
This non-fluorescent, cell-permeable total ROS detection dye reacts directly with a wide range of reactive species, such as hydrogen peroxide, peroxynitrite and hydroxyl radicals, yielding a green fluorescent product indicative of cellular production of different ROS/RNS types.