Order Entry
Canada
ContactUsLinkComponent
TOTAL-NUCLEAR-ID® Green/Red Nucleolar/Nuclear Detection Kit, Enzo Life Sciences
TOTAL-NUCLEAR-ID® Green/Red Nucleolar/Nuclear Detection Kit, Enzo Life Sciences
Catalog # 89165-874
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
TOTAL-NUCLEAR-ID® Green/Red Nucleolar/Nuclear Detection Kit, Enzo Life Sciences
Catalog # 89165-874
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-51006-500
CAS Number:  

Specifications

  • Size:
    1 Kit
  • Storage conditions:
    −80 °C
  • Application:
    Fluorescence Microscopy, Fluorescent Detection
  • Cat. no.:
    89165-874
  • Supplier No.:
    51006-500

Specifications

About this item

The kit contains two proprietary dyes suitable for visualizing nucleoli and nuclei simultaneously in living cells.

The dyes allow examination of nucleolar dynamic changes in intracellular distribution, trafficking and localization arising from biological processes such as the cell cycle and ribosome biogenesis

A control nucleolus perturbation agent, actinomycin D, is provided for monitoring changes in nucleolar dynamics. Potential applications for this kit include monitoring impaired ribosome biogenesis, inhibition transcription, cell cycle dynamics and cellular stress, as well as the distribution, trafficking and dynamics of nucleolar proteins, the distribution of viral proteins, and potentially as an aid in identifying cancer cells.

The kit is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms.

Delivery: Kit contains Nucleolar-ID™ Green Detection Reagent, 50 µL, Nuclear-ID™ Red Detection Reagent, 50 µL, Actinomycin D Control, 125 µg and 10X Assay Buffer, 15 mL.