To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1 mM
- Description:Gold 525 dUTP
- Size:25 nmoL
- Purity detail:Purified by ion-exchange chromatography.
- Purity:≥93% (HPLC)
- Formulation:Liquid. Solution in water.
- Cat. no.:89165-850
- Supplier No.:42843
Specifications
About this item
Gold 525 [5(6)-Carboxyrhodamine 6G] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Gold 525 dUTP can also be used for multicolor fluorescence labelling.