To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Description:Bio-7-dATP
- Size:50 nmoL
- Environmentally Preferable:
- Purity detail:Contains <5% Bio-7-dADP.
- Purity:≥93% (HPLC)
- Concentration:1mM
- Formulation:Liquid. Solution in water.
- Cat. no.:89165-840
- Supplier No.:42819
Bio-7-dATP (Biotin-7-2’-deoxyadenosine-5’-triphosphate) can replace dATP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Bio-7-dATP can be used to produce biotinylated DNA probes in a variety of labelling reactions including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. The resulting biotin-labeled probe can be used in a variety of hybridization applications incuding Southern blots, Northern blots, or dot blots. The probes can also be used for in situ hybridization procedures on fixed cells and tissues. The biotinylated probes have been shown to hybridize to homologous nucleic acid at the same rate and to the same extent as non-biotinylated probes. The hybridized biotinylated DNA probes can be detected by their interaction with biotin-binding proteins, such as avidin, streptavidin or antibodies coupled to fluorescent dyes or color-producing enzymes.