Bio-11-CTP, Enzo Life Sciences
Catalog # 89165-838
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:Bio-11-CTP
- Size:250 nmoL
- Purity detail:Contains <5% Bio-11-CDP.
- Purity:≥93% (HPLC)
- Concentration:10mM
- Formulation:Liquid. Solution in water.
- Cat. no.:89165-838
- Supplier No.:42818
Specifications
About this item
Bio-11-CTP (Biotin-11-cytidine-5’-triphosphate) can replace CTP for in vitro transcription reaction catalysed by T3, T7 or SP6 RNA polymerases.
- Appearance: Clear, colorless liquid
- Purity: ≥93% (HPLC)
- Concentration: 10 mM
- Extinction coefficient: 10,700 M-1 cm-1 (240 nm, pH 7,0)
The biotin-labelled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridisation, sequencing, and genome analysis.
The biotin-labelled hybridised probes can be detected by a reporter molecule linked to streptavidin, avidin, or anit-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble coloured precipitate.