ContactUsLinkComponent
BIO-PROBE® 3’-Oligonucleotide Labeling Kit With Bio-16-ddUTP, Enzo Life Sciences
BIO-PROBE® 3’-Oligonucleotide Labeling Kit With Bio-16-ddUTP, Enzo Life Sciences
Catalog # 89165-812
Supplier:  Enzo Life Sciences
CAS Number:  
BIO-PROBE® 3’-Oligonucleotide Labeling Kit With Bio-16-ddUTP, Enzo Life Sciences
Catalog # 89165-812
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-42730-31
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BIO-PROBE® 3’-Oligonucleotide Labeling Kit With Bio-16-ddUTP
  • Size:
    1 Kit
  • Cat. no.:
    89165-812
  • Supplier No.:
    42730-31
Specifications
About this item
Terminal labeling is an ideal procedure for the labeling of oligonucleotides with haptens such as biotin or fluorescein.
An oligonucleotide can be synthesized using standard, commercially available reagents and labeled after synthesis in a simple and reproducible enzyme reaction.The BioProbe® 3'-Oligonucleotide Labeling Kit with Bio-16-ddUTP contains all reagents required for terminal labeling of oligonucleotides with biotin except the oligonucleotides to be labeled.The BioProbe® 3'-Oligonucleotide Labeling Kit provides maximum flexibility for labeling oligonucleotides approximately 15 to 100 nucleotides in length. A standard reaction will label up to 100 picomoles (1 µg of a 30-mer oligonucleotide sequence).
A complete kit contains two component packs that are available separately: a Reagent Pack and a Bio-16-ddUTP Dideoxynucleotide Pack. Both packs contain sufficient reagents for 25 labeling reactions.
Ordering information: Contents: BioProbe® 3'-Oligonucleotide Labeling System Reagent Pack (Prod. No. ENZ-42730)Bio-16-ddUTP Deoxynucleotide Pack for 3'-Oligonucleotide Labeling (Prod. No. ENZ-42731)