Order Entry
Canada
ContactUsLinkComponent
Wash Buffer Salts (3 packs), Enzo Life Sciences
Wash Buffer Salts (3 packs), Enzo Life Sciences
Catalog # 89165-714
Supplier:  Enzo Life Sciences
CAS Number:  
Wash Buffer Salts (3 packs), Enzo Life Sciences
Catalog # 89165-714
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-33802
CAS Number:  

Specifications

  • Size:
    3 L
  • Formulation:
    Lyophilized.
  • Sensitivity:
    Dissolve entire contents of packet in 1 liter of distilled or deionized water. The pH of the solution should be 7.1 ± 0.1.
  • Cat. no.:
    89165-714
  • Supplier No.:
    33802

Specifications

About this item

Wash Buffer Salts can be used in tissue preparations, immunohistochemical staining procedures and in assays that require HRP-linked detection reagents

Each packet of salts yields 1 liter of buffered saline.