To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:50 µg
- Protein synonyms:UB2
- Protein/peptide name:Di-Ubiqutin (K63 linked)
- Purity:>95% (SDS-PAGE)
- Concentration:2.5mg/ml
- Formulation:Liquid. In 50mM TRIS, pH 7.6, containing 1mM EDTA.
- Cat. no.:89165-206
- Supplier No.:UW0730-0050
Specifications
About this item
Protein components produced in E. coli. Synthesised enzymatically in vitro.
Polyubiquitin chains, linked through specific lysine residues, are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.