HL60 Cell Extract (Non-induced), Enzo Life Sciences
Catalog # 89164-902
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:Immunoblotting Standard, HL60 Cell Extract (Non-Induced)
- Size:200 µl
- Cat. no.:89164-902
- Supplier No.:SW101-0200
Specifications
About this item
Whole-cell extract of human HL-60 leukemia cells.
- Immunoblotting standard
- Supplied in SDS-PAGE sample buffer
- Ready-to-use
WB control for PARP antibodies, to detect intact 116 kDa PARP. Recommended gel load is 20 µl per lane (~75,000 cells).