Order Entry
ContactUsLinkComponent
Human Recombinant HDAC11 (from Insect cells (Sf9)), HIS Tag
Human Recombinant HDAC11 (from Insect cells (Sf9)), HIS Tag
Catalog #: 89164-534
Supplier:  Enzo Life Sciences
CAS Number:  
HISTONE DEACETYLASE 11 REC 50UG
Human Recombinant HDAC11 (from Insect cells (Sf9)), HIS Tag
Catalog #: 89164-534
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    HIS Tag
  • Biological activity:
    Hydrolysis of N-ε-acetyl-lysine residues of histones.
    >3U/μg. One unit is defined as the amount of enzyme that deacetylates 1 pmol/min of Fluor de Lys™-SIRT1 (Prod. No. BML-KI177) substrate at 37°C / 500μM.
  • Protein/peptide name:
    HDAC11
  • Purity:
    >50% (SDS-PAGE)
  • Formulation:
    Liquid. In 50mM TRIS, pH 8.0, containing 138mM NaCl and 10% glycerol.
  • Cat. no.:
    89164-534
  • Supplier No.:
    SE560-0050

Specifications

About this item

Highly active

Produced in Sf9 cells. HDAC11 from human cDNA, transcript variant 1 (identical to GenBank accession NM_024827) is fused at the N-terminus to a His-tag.

Deacetylates lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events.

Customers Who Bought This Also Bought