Human Recombinant LSD1 (from E. coli)
Catalog #: 89164-494
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Biological activity:>20 U/µg. One U=1 pmol/min at 30°C, 100 µM Histone H3 Dimethyl Lysine-4 Peptide (Prod. No. BML-P256).
- Protein/peptide name:LSD1
- Purity:>90% (SDS-PAGE)
- Formulation:Liquid. Solution containing 6mM Na2HPO4, 1.1mM K2HPO4, pH 7.2, 82mM sodium chloride, 1.6mM KCl and 40% (v/v) glycerol.
- Cat. no.:89164-494
- Supplier No.:SE544-0050
Specifications
About this item
Highly active
Produced in E. coli.
Customers Who Bought This Also Bought