Order Entry
Canada
ContactUsLinkComponent
Human Aminopeptidase N (from Serum)
Human Aminopeptidase N (from Serum)
Catalog # 89164-490
Supplier:  Enzo Life Sciences
CAS Number:  
Human Aminopeptidase N (from Serum)
Catalog # 89164-490
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SE542-0010
CAS Number:  

Specifications

  • Source:
    Serum
  • Species:
    Human
  • Size:
    10 µg
  • Enzyme name:
    Aminopeptidase N
  • Purity:
    Purified
  • Cat. no.:
    89164-490
  • Supplier No.:
    SE542-0010

Specifications

About this item

Isolated from human serum.