Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Tyrosinase (from E. coli)
Human Recombinant Tyrosinase (from E. coli)
Catalog # 89164-476
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant Tyrosinase (from E. coli)
Catalog # 89164-476
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SE535-0100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Enzyme name:
    Tyrosinase
  • Purity:
    ~90% (SDS-PAGE)
  • Cat. no.:
    89164-476
  • Supplier No.:
    SE535-0100

Specifications

About this item

Produced in E. coli.

Tyrosinase is a copper-containing enzyme that catalyzes four steps in the melanin and catecholamine synthesis pathways. Like tyrosine hydroxylase (EC 1.13.16.2), it can catalyze the formation of L-DOPA in the brain and nervous system. It oxidizes monophenols, but like catecholoxidase (EC 1.10.3.1), it can also oxidize o-diphenols to o-quinones. It is involved in cancer, Parkinson’s, pigmentation diseases, Alzheimer’s and obesity. Much of the research and inhibitor screening has used mushroom tyrosinase. However, mushroom tyrosinase interacts very differently with substrates and inhibitors compared to the human enzyme. In addition, commercially available mushroom tyrosinase is not always of good purity. Recombinant versions of human tyrosinase amino acids 1-345 or 1-378, also available commercially, are not functional due the lack of critical catalytic and structural residues.