Order Entry
Canada
ContactUsLinkComponent
Human Recombinant VHR dual specificity phosphatase (from E. coli)
Human Recombinant VHR dual specificity phosphatase (from E. coli)
Catalog # 89164-022
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant VHR dual specificity phosphatase (from E. coli)
Catalog # 89164-022
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SE333-0050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Enzyme name:
    VHR dual specificity phosphatase
  • Cat. no.:
    89164-022
  • Supplier No.:
    SE333-0050

Specifications

About this item

Produced in E. coli.