Human Recombinant Caspase-3 (from E. coli)
Catalog # 89163-832
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:5000 U
- Enzyme name:Caspase-3
- Enzyme activity:8000 U/µg. One U=1 pmol/min using Ac-DEVD-pNA (200 µM; Prod. No. BML-P412) as substrate at 30°C
- Purity:≥95% (SDS-PAGE)
- Formulation:100 U/µl in 50 mM HEPES, pH 7.4, 100 mM sodium chloride, 0.5% CHAPS, 1 mM EDTA, 10% glycerol and 10 mM dithiothreitol.
- Cat. no.:89163-832
- Supplier No.:SE169-5000
Specifications
About this item
Produced in E. coli. The enzyme is cleaved and activated from the proenzyme.
Caspase-3 (CPP32; Yama; apopain) from human cDNA, expressed in an E. coli expression system. The enzyme was cleaved and activated from the proenzyme. Can be used to study enzyme regulation and kinetics, cleave target substrates and screen for inhibitors.