Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Casein kinase II (from E. coli)
Human Recombinant Casein kinase II (from E. coli)
Catalog # 89163-786
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant Casein kinase II (from E. coli)
Catalog # 89163-786
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SE124-0010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 KU
  • Form:
    Liquid
  • Enzyme name:
    Casein kinase II
  • Enzyme activity:
    700 kU/mg protein. One unit will transfer 1 pmol phosphate from ATP to peptide substrate per minute at 30 °C
  • Purity:
    Free from proteases and phosphatases.
  • Concentration:
    ~0.7 mg/ml
  • Formulation:
    In 20 mM TRIS-HCl, pH 7.5, containing 0.35 M sodium chloride, 1 mM EDTA, 2 mM DTT and 0.1% Triton X-100.
  • Cat. no.:
    89163-786
  • Supplier No.:
    SE124-0010

Specifications

About this item

Produced in E. coli. Consists of 44kDa alpha catalytic and 26kDa ß regulatory subunits as an alpha2ß2 tetramer.