Human Recombinant T cell protein tyrosine phosphatase (from E. coli)
Catalog # 89163-776
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:200 U
- Form:Liquid
- Enzyme name:T cell protein tyrosine phosphatase
- Purity:Contains no detectable protease, DNase, RNase, or Ser/Thr phosphatase activity.
- Formulation:In 30% glycerol containing stabilizers.
- Cat. no.:89163-776
- Supplier No.:SE114-0200
Specifications
About this item
Produced in E. coli. T-cell phosphatase with a C-terminal 11 kDa deletion (TCdeltaC11) (aa 1-317).