Staphylococcus aureus Sphingomyelinase
Supplier: Enzo Life Sciences
Certificates
About this item
Isolated from Staphylococcus aureus.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Species:Staphylococcus aureus
- Form:Liquid
- Enzyme name:Sphingomyelinase
- Enzyme activity:Specific activity: 100 - 300 U/mg protein. One unit will hydrolyze 1 µmol of TNPAL-sphingomyelin per minute at pH 7.4 and 37 °C
- Formulation:Supplied in 50% glycerol/0.25 M phosphate buffer.
Specifications
Frequently Bought Together