About this item
MMP inhibitor
STTHWGFTLS is ten times less potent than, thus a negative control for CTTHWGFTLC (Prod. No. BML-PI136), which is a specific inhibitor of the gelatinases MMP-2 and MMP-9. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo.