N-Acetyl-Leu-Leu-Methiona
Supplier: Enzo Life Sciences
Certificates
About this item
Calpain inhibitor
Cell-permeable, peptide aldehyde inhibitor of calpain and other neutral cysteine proteases. Potent inhibitor of cathepsin L (Ki=0.6nM) and cathepsin B (Ki=100nM).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.