Specifications
- Size:1 mg
- Protein/peptide name:Egfr (988-998), Ptp1B Substrate
- Cat. no.:89161-232
- Supplier No.:P323-0001
Specifications
About this item
PTP1B substrate
This sequence, DADEpYLIPQQG, is from an autophosphorylation site (Tyr992) of epidermal growth factor receptor (EGFR). An excellent substrate for mammalian PTP1B with Km=3.9 µM ) and Yersinia PTPase. May be used in non-radioactive phosphatase assays incorporating the BIOMOL GREENTM reagent.