Specifications
- Size:1 mg
- Protein/peptide name:YEEI Peptide
- Purity:>98%
- Cat. no.:89161-194
- Supplier No.:P300-0001
Specifications
About this item
Src kinase activator
Sequence is from hamster polyoma virus middle T inclusive of Tyr324. This 11-residue phosphopeptide activates Src family members by binding with high affinity (Kd=3-6 nM for Src SH2) to their SH2 domains. The crystal structures of this peptide complexed to Src and Lck SH2 domains have been determined.