To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Biotin
- Size:1 mg
- Protein/peptide name:SAMS Peptide
- Purity:95%
- Cat. no.:89161-118
- Supplier No.:P232-0001
Specifications
About this item
AMPK substrate
This peptide, biotin-HMRSAMSGLHLVKRR, contains a biotin group at its amino terminus. It is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.