To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:5 mg
- Protein/peptide name:Meosuc-Aapv-Amc (Fluorogenic Substrate)
- Purity:98%
- Cat. no.:89161-102
- Supplier No.:P224-0005
Specifications
About this item
Elastase substrate
Highly sensitive substrate for neutrophil elastase (leukocyte elastase) (Km=130 µM), but not granzyme H. This substrate can be used for in vitro assays with purified enzyme, on live cells, or in conditioned medium. Ex: 380 nm, Em: 460 nm.