Specifications
- Size:1 mg
- Protein/peptide name:Abl Kinase Substrate
- Purity:95%
- Cat. no.:89161-096
- Supplier No.:P216-0001
Specifications
About this item
This peptide, EAIYAAPFAKKK, was identified as a high affinity (Km=4 µM) and specific substrate for the Abl kinase through a degenerate peptide library. The three lysines on the carboxy-terminus allow this peptide to be used in phosphocellulose kinase assays.