Specifications
- Size:1 mg
- Protein/peptide name:Fyn Kinase Substrate
- Purity:95%
- Cat. no.:89161-094
- Supplier No.:P215-0001
Specifications
About this item
This peptide, EFGTYGTLSKKK, is derived from a peptide (pF4) discovered, through phage display, to be a very good substrate for Fyn kinase. Its Km is estimated to be 70 µM. The addition of three lysines on the C-terminus allows this peptide to be used in phosphocellulose kinase assays.