Specifications
- Size:0.5 mg
- Protein/peptide name:Cam Kinase II
- Purity:97%
- Cat. no.:89161-064
- Supplier No.:P200-0500
Specifications
About this item
CaM kinase inhibitor
Synthetic peptide with sequence from Ca2+/calmodulin-dependent protein kinase II pseudosubstrate site. Peptide binds calmodulin tightly and inhibits calmodulin-requiring enzymes including CaMKII, phosphodiesterase and calcineurin. IC50=52 nM for CaMKII in the presence of 100 nM calmodulin.