Specifications
- Size:1 mg
- Protein/peptide name:MAP Kinase (177-189) Phosphorylated
- Cat. no.:89161-040
- Supplier No.:P164-0001
Specifications
About this item
VHR substrate
Diphosphopeptide with sequence, DHTGFLpTEpYVATR, corresponding to MAP kinase residues 177-189 inclusive of putative activation sites. Peptide can be used as a VHR phosphatase substrate (BML-SE333). Phosphate release may be quantified using BML-AK111 malachite green colorimetric reagent. Phosphate release is much more rapid for the phosphotyrosine residue compared to the phosphothreonine residue, both in vitro and in vivo.