Order Entry
ContactUsLinkComponent
Abz-Frk(Dnp)-P
Abz-Frk(Dnp)-P
Catalog #: 89161-034
Supplier:  Enzo Life Sciences
CAS Number:  
ABZ FRKDNP P
Abz-Frk(Dnp)-P
Catalog #: 89161-034
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 mg
  • Protein/peptide name:
    Abz-Frk(Dnp)-P
  • Purity:
    95%
  • Cat. no.:
    89161-034
  • Supplier No.:
    P161-0001

Specifications

About this item

ACE substrate

Fluorogenic substrate for angiotensin converting enzyme-1 (ACE1). The somatic form of ACE1 is a zinc-dependent peptidyl dipeptidase comprising two catalytic domains referred to as the N- and C-domains. The testicular form, due to a downstream start site, contains only the C-domain. Abz fluorescence is quenched by the Dnp group until cleavage (at Arg-Lys) separates them. Ex: 320 nm, Em: 420 nm. This quenched substrate is useful for assay of plasma, tissue, and cell culture ACE1 activity, inhibitor screening and kinetic analysis.

Customers Who Bought This Also Bought