Specifications
- Size:1 mg
- Protein/peptide name:Crosstide
- Cat. no.:89161-012
- Supplier No.:P149-0001
Specifications
About this item
Akt substrate
This peptide sequence, GRPRTSSFAEG, corresponds to the sequence of GSK3 surrounding the serine phosphorylated by MAP kinase activated protein kinase (MAPKAP kinase-1) and p70 ribosomal S6 kinase. Acts as a substrate for Akt (PKB), Km=4µM. Useful in phosphocellulose kinase assays.