Specifications
- Size:25 mg
- Protein/peptide name:Suc-Ala-Ala-Pro-Phe-pNA
- Purity:99%
- Cat. no.:89160-996
- Supplier No.:P141-0025
Specifications
About this item
Cathepsin substrate
Chromogenic substrate cleaved by cathepsin G, subtilisins, chymotrypsin, chymase, and cyclophilins, but not neutrophil elastase. Release of pNA is monitored at 405-410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145).