To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 mg
- Protein/peptide name:Fluorescent Substrate
- Purity:98%
- Cat. no.:89160-966
- Supplier No.:P126-0001
Specifications
About this item
Highly quenched and sensitive fluorogenic MMP substrate.
Fluorogenic substrate for nearly all matrix metalloproteinase (MMP) enzymes, and ADAM17/TACE. This highly-quenched (99.5%) substrate is useful for inhibitor screening and kinetic analysis. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have been used: 320 – 340/400 – 420. Also available: OmniMMPTM Fluorogenic Control Peptide (BML-P127).