Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
EGTA (2X) Buffer, Enzo Life Sciences
EGTA (2X) Buffer, Enzo Life Sciences
EGTA (2X) Buffer, Enzo Life Sciences
Catalog # 89160-312
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
EGTA (2X) Buffer, Enzo Life Sciences
Catalog # 89160-312
Supplier:
Enzo Life Sciences
Supplier Number:
BML-KI136-0001
CAS Number:
Specifications
Size:
1 mL
Cat. no.:
89160-312
Supplier No.:
KI136-0001
Specifications
About this item
Product Details
Product Details