FLUOR DE LYS® Developer I Concentrate, Enzo Life Sciences
Catalog # 89160-284
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:Fluor-de-Lys® Developer I concentrate
- Size:300 µl
- Concentration:20X concentrate after dilution (1X)
- Cat. no.:89160-284
- Supplier No.:KI105-0300
- Sufficient For:100 assays (50 µl reaction volume)
Specifications
About this item
FLUOR DE LYS® Developer is an essential reagent for signal generation in HDAC or sirtuin assays done with the FLUOR DE LYS® substrate. An appropriate FLUOR DE LYS® family substrate, comprising an acetylated lysine side chain, is first incubated with a sample containing HDAC activity (nuclear or cellular extract, purified enzyme, bead-bound immunocomplex, etc.).
- An essential reagent for signal generation in HDAC or sirtuin assays done with the FLUOR DE LYS® substrate.
Deacetylation of the substrate sensitizes it so that, in a second step, treatment with the FLUOR DE LYS® Developer produces a fluorophore. Typically, an 'inhibitor stop' will be included in the 1x Developer dilution (e.g. Trichostatin A) for class I and II HDACs, nicotinamide for sirtuins).