To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:140 µg
- Protein/peptide name:Dendrotoxin-I (Dendroaspis P. Polylepis)
- Purity:>98% (HPLC)
- Formulation:Lyophilized
- Cat. no.:89160-252
- Supplier No.:KC163-0140
Specifications
About this item
Ultra-pure
K+ channel blocker. Blocks Kv1.2 and Kv1.1 channels (IC50=0.13 and 3.1nM in oocytes respectively, with higher values for mammalian cells), as well as heteromultimeric channels comprised of Kv1.2 and Kv1.1 with other Kv1 isoforms.