Specifications
- Size:100 µg
- Protein/peptide name:Blocking Peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
- Purity:>96%
- Cat. no.:89160-114
- Supplier No.:HP9301-0100
Specifications
About this item
Synthetic peptide corresponding to amino acid residues [12-25] + Cys-NH2 of human heme oxygenase-1. Complementary peptide to rabbit polyclonal antibody (BML-HC3001) for use in control experiments. Preincubation of antibody with this peptide abolishes the antibody reactivity with HO-1 protein.