Order Entry
Canada
Orders LinkContactUsLinkComponent
AUDA ≥98% (by TLC)

AUDA ≥98% (by TLC)

Supplier:   Enzo Life Sciences
AUDA ≥98% (by TLC)
Supplier:  Enzo Life Sciences
Certificates
AUDA ≥98% (by TLC)
AUDA ≥98% (by TLC)

1 Options Available Below

Product Details & Documents

Novel inhibitor of soluble epoxide hydrolase (IC50=18nM and 69nM for the mouse and human enzymes respectively). Produces a hypotensive effect in angiotensin-infused rats and prevents and reverses cardiac hypertrophy in mouse models of cardiac hypertrophy.

  • Epoxide hydrolase inhibitor

White solid.

Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml).

Synonyms:  12-(3-Adamantan-1-yl-ureido)-dodecanoic acid, 12-(3-(Adamantan-1-yl)ureido)dodecanoic acid
  • CAS Number:479413-70-2
  • Formula:C₂₁H₃₆N₂O₃
  • MDL Number:MFCD12912267
  • Molecular Weight:392.575 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 mg
    Discontinued
    Supplier Number: -
  • 50 mg
    Log in for availability
    Log in for availability
    Each
    Each - $583.03
    $583.03
    Each
       $583.03
    Minimum order quantity is 1

    Supplier Number: BML-EI392-0050
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.